Please login to your online account to display your discounted pricing

ICI 215,001 hydrochloride, Tocris

β3 agonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ICI 215, 001 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H23NO6·HCl
Formula Weight 397.86
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

092950

 
r & d systems
RDS092950
50mg Each for $746.48

092910

 
r & d systems
RDS092910
10mg Each for $177.37
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 397.86
CAS 140850-02-8
Chemical Name or Material ICI 215, 001 hydrochloride
Molecular Formula C19H23NO6·HCl
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
Safety and Handling

Store at Room Temperature