ICI 215,001 hydrochloride, Tocris
β3 agonist
Manufacturer:  R & D Systems RDS092950
 View more versions of this product
Catalog No. 092950
Specifications Description & Specifications
Specifications
ICI 215, 001 hydrochloride
140850-02-8
C19H23NO6·HCl
(S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
50mg
>98%
397.86
Chemical Identifiers
>98%
397.86
ICI 215, 001 hydrochloride
C19H23NO6·HCl
(S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
Safety and Handling
Recommended Storage : Store at Room Temperature