Please login to your online account to display your discounted pricing

ICI 199,441 hydrochloride, Tocris

Potent κ agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material ICI 199, 441 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H24Cl2N2O·HCl
Formula Weight 427.8
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

077850

 
r & d systems
077850
50mg Each for $762.58

077810

 
r & d systems
077810
10mg Each for $181.19
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 427.8
Chemical Name or Material ICI 199, 441 hydrochloride
Assay Percent Range >99%
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Molecular Formula C21H24Cl2N2O·HCl
CAS 115199-84-3
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature