Please login to your online account to display your discounted pricing

ICI 199,441 hydrochloride, Tocris


Potent κ agonist

Quantity 50mg

Manufacturer: r & d systems  077850

 View more versions of this product

Catalog No. 077850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 427.8
Assay Percent Range >99%
Chemical Name or Material ICI 199, 441 hydrochloride
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Molecular Formula C21H24Cl2N2O·HCl
CAS 115199-84-3
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material ICI 199, 441 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H24Cl2N2O·HCl
Formula Weight 427.8
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Accessories

Store at Room Temperature