missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ICI 192,605, Tocris

Potent thromboxane A2/TP receptor antagonist

$176.10 - $741.11

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C22H23ClO5
Formula Weight 402.87
Synonym 4-(Z)-6-(2-o-Chlorophenyl-4-o-hydroxyphenyl-1, 3-dioxan-cis-5-yl)hexenoic acid
Chemical Name or Material ICI 192, 605
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
083710 R & D Systems
RDS083710
10mg Each for $176.10
083750 R & D Systems
RDS083750
50mg Each for $741.11
Specifications Description & Specifications

Chemical Identifiers

>98%
402.87
ICI 192, 605
C22H23ClO5
4-(Z)-6-(2-o-Chlorophenyl-4-o-hydroxyphenyl-1, 3-dioxan-cis-5-yl)hexenoic acid

Specifications

10mg
>98%
402.87
ICI 192, 605
117621-64-4
C22H23ClO5
4-(Z)-6-(2-o-Chlorophenyl-4-o-hydroxyphenyl-1, 3-dioxan-cis-5-yl)hexenoic acid

Safety and Handling

Recommended Storage : Desiccate at -20°C