Please login to your online account to display your discounted pricing

ICI 185,282, Tocris


Potent thromboxane receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS083610

 View more versions of this product

Catalog No. 083610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 374.36
Chemical Name or Material ICI 185, 282
Assay Percent Range >98%
Molecular Formula C18H21F3O5
CAS 106393-80-0
Synonym 5-(Z)-7-(4-o-Hydroxyphenyl-2-trifluoromethyl-1, 3 dioxan-cis-5-yl)heptenoic acid
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material ICI 185, 282
Assay Percent Range >98%
Molecular Formula C18H21F3O5
Formula Weight 374.36
Synonym 5-(Z)-7-(4-o-Hydroxyphenyl-2-trifluoromethyl-1, 3 dioxan-cis-5-yl)heptenoic acid
Accessories

Desiccate at -20°C