Please login to your online account to display your discounted pricing

ICI 185,282, Tocris

Potent thromboxane receptor antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ICI 185, 282
Assay Percent Range >98%
Molecular Formula C18H21F3O5
Formula Weight 374.36
Synonym 5-(Z)-7-(4-o-Hydroxyphenyl-2-trifluoromethyl-1, 3 dioxan-cis-5-yl)heptenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

083650

 
r & d systems
RDS083650
50mg Each for $746.48

083610

 
r & d systems
RDS083610
10mg Each for $177.37
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 374.36
Chemical Name or Material ICI 185, 282
Assay Percent Range >98%
Molecular Formula C18H21F3O5
CAS 106393-80-0
Synonym 5-(Z)-7-(4-o-Hydroxyphenyl-2-trifluoromethyl-1, 3 dioxan-cis-5-yl)heptenoic acid
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C