Please login to your online account to display your discounted pricing

ICI 182,780, Tocris


Estrogen receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS104710

 View more versions of this product

Catalog No. 104710

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 606.77
Quantity 10mg
Chemical Name or Material ICI 182, 780
Assay Percent Range >99%
Molecular Formula C32H47F5O3S
CAS 129453-61-8
Synonym 7α,17β-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material ICI 182, 780
Assay Percent Range >99%
Molecular Formula C32H47F5O3S
Formula Weight 606.77
Synonym 7α,17β-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol
Accessories

Store at Room Temperature