Please login to your online account to display your discounted pricing

Corning™ Ice Pan, Mini 1L


Multi-purpose EVA pans that provide a non-toxic alternative to conventional ice pans

Manufacturer: corning™  C432119

 View more versions of this product

Catalog No. 07-210-119

  • / Case
 

Description & Specifications

Specifications

Quantity 1 per case
Capacity 1L
Color Lime Green
Dimensions (L x W x H) 15.9 x 15.9 x 9.5cm