Please login to your online account to display your discounted pricing
Thermo Scientific iCE™ 3300 AAS Atomic Absorption Spectrometer
For reliable, accurate, ultra-flexible elemental analysis and superb value for money the iCE™ 3300 Flame or Furnace AAS from Thermo Scientific™ delivers.
Products
Catalog Number Mfr. No. Item Description Price Quantity    
14-386-471
thermo scientific
942350033302
iCE 3300 Double Beam AA Spectrometer with 100mm Universal Burner CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Here is a compact, AA system which offers a simple and reliable solution for trace elemental analysis; the Thermo Scientific™ iCE™ 3300 AAS is designed for ultimate ease-of-use and offers superb value for money.
With options to configure the instrument as a dedicated flame, dedicated furnace or flame and furnace system, this is a truly versatile analyzer. Robust enough for samples with high levels of dissolved solids or acids, all users will appreciate the sensitivity, accuracy and uncomplicated approach of the iCE 3300 AAS.
Three Options to Suit your Laboratory’s Needs
Dedicated Flame
A single flame atomizer AAS with a fully automatic gas box, the iCE3300 Flame system delivers a complete solution for laboratories with a primary requirement for flame analysis.
Dedicated Furnace
The low cost iCE3300 Graphite Furnace system provides a complete solution for laboratories with a main need for elemental analysis by furnace.
Dual Flame and Furnace
Simply add the optional Thermo Scientific™ GFS33™ Graphite Furnace and Autosampler to your iCE3300 Flame AAS instrument to create a compact, interchangeable, multi atomizer and access the best of both worlds.
Maximize Productivity
  • Increase your sample through-put and limit downtime with productivity enhancing technology.
  • Optimization routines can be included in the method to ensure truly optimal parameters
  • Thermo Scientific™ SOLAAR™ software offers ultimate ease-of-use with extensive help functions, automated performance controls and wizards
  • Self-calibrating monochromators and auto-aligning lamps offer simple optical setup and optimum light throughput in dedicated Flame system
  • Parts-per-billion ultra-trace detection capability from dedicated Graphite Furnace system
Gain Cost Efficiency
  • Gain even greater efficiency from hardware and accessories designed to maximize cost efficiency.
  • Fully automatic gas control system ensures repeatable flame conditions and safety
  • Configure the fully customizable auto-sampler to maximize through-put
  • Designed to ensure lowest possible gas flows to significantly reduce cost of consumables
  • Innovative Extended Lifetime Cuvettes (ELC) for reduced consumable costs, can be used for thousands of Graphite Furnace firings
Enjoy Advanced Performance
Improve your analysis with high performing instruments the deliver accuracy and precision.
  • Unbeatable Flame performance with superior detection limits and a fully inert sample system
  • Graphite Furnace obtains drift free results from automated cuvette heating control meaning, for accurate, repeatable analysis
  • Guaranteed Deuterium background correction for high accuracy
  • Flame option uses a universal finned titanium burner to enable the use of both air/acetylene and nitrous oxide/acetylene flame types and maintains a stable, reproducible flame
  • Graphite Furnace TV (GFTV) real time viewing enables precise analysis, time after time