missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Icaritin 96.0+%, TCI America™
$708.33 - $708.33
Chemical Identifiers
| CAS | 118525-40-9 |
|---|---|
| Molecular Formula | C21H20O6 |
| Molecular Weight (g/mol) | 368.39 |
| MDL Number | MFCD22422519 |
| InChI Key | TUUXBSASAQJECY-UHFFFAOYSA-N |
| Synonym | 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one |
| PubChem CID | 5318980 |
| IUPAC Name | 3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one |
| SMILES | COC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C(CC=C(C)C)=C2O1 |
Chemical Identifiers
| 118525-40-9 | |
| 368.39 | |
| TUUXBSASAQJECY-UHFFFAOYSA-N | |
| 5318980 | |
| COC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C(CC=C(C)C)=C2O1 |
| C21H20O6 | |
| MFCD22422519 | |
| 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one | |
| 3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one |
Specifications
| 118525-40-9 | |
| C21H20O6 | |
| 50 mg | |
| TUUXBSASAQJECY-UHFFFAOYSA-N | |
| 3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one | |
| 5318980 | |
| ≥96.0% (HPLC) | |
| Icaritin |
| 233°C | |
| MFCD22422519 | |
| 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one | |
| COC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C(CC=C(C)C)=C2O1 | |
| 368.39 | |
| 368.39 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DJ3100870
TSCA : No
Recommended Storage : Refrigerator