missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Icariin, 98%

$186.59 - $675.45

Chemical Identifiers

CAS 489-32-7
Molecular Formula C33H40O15
Molecular Weight (g/mol) 676.668
MDL Number MFCD00210516
InChI Key TZJALUIVHRYQQB-JOJMVVPMSA-N
Synonym 3-[(6-Deoxy-alpha-L-mannopyranosyl)oxy]-7-beta-D-glucopyranosyloxy-5-hydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one
PubChem CID 131801625
IUPAC Name 5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66324MD View Documents Alfa Aesar™
J66324MD
250mg Each for $186.59
 
AAJ6632403 View Documents Alfa Aesar™
J6632403
1g Each for $675.45
 
Specifications

Chemical Identifiers

489-32-7
676.668
TZJALUIVHRYQQB-JOJMVVPMSA-N
131801625
CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O
C33H40O15
MFCD00210516
3-[(6-Deoxy-alpha-L-mannopyranosyl)oxy]-7-beta-D-glucopyranosyloxy-5-hydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one
5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one

Specifications

Yellow
489-32-7
MFCD00210516
3-[(6-Deoxy-alpha-L-mannopyranosyl)oxy]-7-beta-D-glucopyranosyloxy-5-hydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one
CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O
676.668
676.66
98%
250mg
C33H40O15
14,3617
TZJALUIVHRYQQB-JOJMVVPMSA-N
5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
131801625
Powder
Icariin
Safety and Handling

Safety and Handling

RTECSNumber : DJ2980500