Please login to your online account to display your discounted pricing

ICA 110381, Tocris™

KV7.2/7.3 activator; displays anticonvulsant properties

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material ICA 110381
Assay Percent Range >98%
Molecular Formula C12H8Cl2N2O
Formula Weight 267.11
Synonym 4-Chloro-N-(6-chloro-3-pyridinyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

495010

 
r & d systems
RDS495010
10mg Each for $177.37

495050

 
r & d systems
RDS495050
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 267.11
Quantity 10mg
Chemical Name or Material ICA 110381
Assay Percent Range >98%
Molecular Formula C12H8Cl2N2O
CAS 325457-99-6
Synonym 4-Chloro-N-(6-chloro-3-pyridinyl)benzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C