Please login to your online account to display your discounted pricing

ICA 069673, Tocris™

KV7.2/KV7.3 channel opener

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material ICA 069673
Assay Percent Range >99%
Molecular Formula C11H6ClF2N3O
Formula Weight 269.63
Synonym N-(2-Chloro-5-pyrimidinyl)-3, 4-difluorobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

430510

 
r & d systems
430510
10mg Each for $214.12

430550

 
r & d systems
430550
50mg Each for $891.94
Description & Specifications

Specifications

Formula Weight 269.63
Quantity 10mg
CAS 582323-16-8
Synonym N-(2-Chloro-5-pyrimidinyl)-3, 4-difluorobenzamide
Chemical Name or Material ICA 069673
Molecular Formula C11H6ClF2N3O
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Safety and Handling

Store at Room Temperature