missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ibutilide hemifumarate salt, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H56358MB
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ibutilide hemifumarate salt | |
C44H76N4O10S2 | |
25 mg | |
ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate | |
OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 | |
885.23 | |
442.61 |
122647-32-9 | |
MFCD01715410 | |
14,4883 | |
PCIOHQNIRPWFMV-WXXKFALUNA-N | |
(E)-but-2-enedioic acid;N-[4-[4-[ethyl(heptyl)amino]-1-hydroxybutyl]phenyl]methanesulfonamide | |
5281065 | |
99% |
Chemical Identifiers
122647-32-9 | |
885.23 | |
PCIOHQNIRPWFMV-WXXKFALUNA-N | |
5281065 | |
OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 |
C44H76N4O10S2 | |
MFCD01715410 | |
ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate | |
(E)-but-2-enedioic acid;N-[4-[4-[ethyl(heptyl)amino]-1-hydroxybutyl]phenyl]methanesulfonamide |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : PB0475700
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only