Please login to your online account to display your discounted pricing

Ibuprofen, USP grade, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0219021605

 View more versions of this product

Catalog No. ICN19021605

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Ibuprofen
CAS 15687-27-1
Purity Grade USP
Physical Form White powder
Molecular Formula C13H18O2
Formula Weight 206.3g/mol
MDL Number MFCD00010393
Synonym α-Methyl-4-[2-methylpropyl] benzeneacetic acid
Recommended Storage Store at Room Temperature (15° to 30°C)
EINECS Number 239-784-6

Specifications

Chemical Name or Material Ibuprofen
CAS 15687-27-1
Purity Grade USP
Physical Form White powder
Molecular Formula C13H18O2
Formula Weight 206.3g/mol
MDL Number MFCD00010393
Synonym α-Methyl-4-[2-methylpropyl] benzeneacetic acid
Accessories

Store at Room Temperature (15° to 30°C)