missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(±)-Ibotenic acid monohydrate, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC329130100
Specifications
(±)-Ibotenic acid monohydrate | |
conforms to structure | |
141.0°C to 147.0°C | |
60573-88-8 | |
C5H6N2O4·H2O | |
14, 4877 | |
Solubility in water: soluble in water. | |
C1=C(ONC1=O)C(C(=O)O)N.O | |
176.1 | |
176.1 | |
95% |
Authentic | |
White to Yellow | |
10mg | |
94% min. (HPLC) | |
MFCD00149579 | |
alpha-amino-3-hydroxy-5-isoxazoleacetic acid hydrate, alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat, alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat german, amino-3-hydroxy-5-isoxazolyl acetic acid, ibotenic acid hydrate, ibotenic acid monohydrate, ibotensaeure, ibotensaeure german, isotenic acid, pramuscimol | |
QVKQQLYSTODTJN-UHFFFAOYSA-N | |
2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid;hydrate | |
43362 | |
Powder or Solid |
Chemical Identifiers
60573-88-8 | |
176.1 | |
QVKQQLYSTODTJN-UHFFFAOYSA-N | |
43362 | |
C1=C(ONC1=O)C(C(=O)O)N.O |
C5H6N2O4·H2O | |
MFCD00149579 | |
alpha-amino-3-hydroxy-5-isoxazoleacetic acid hydrate, alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat, alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat german, amino-3-hydroxy-5-isoxazolyl acetic acid, ibotenic acid hydrate, ibotenic acid monohydrate, ibotensaeure, ibotensaeure german, isotenic acid, pramuscimol | |
2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid;hydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger