Please login to your online account to display your discounted pricing

(±)-Ibotenic acid monohydrate, 95%, ACROS Organics™


Quantity 1mg

Manufacturer: thermo fisher scientific  329130010

 View more versions of this product

Catalog No. AC329130010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (HPLC)
Physical Form Powder or Solid
Formula Weight 176.1
Quantity 1mg
Color White to off-white to light yellow
Merck Index 14, 4877
Melting Point 141° to 147°C
Synonym (#1)-α-Amino-3-hydroxy-5-isoxazoleacetic acid monohydrate
Assay Percent Range 95%
CAS 60573-88-8
Chemical Name or Material (+/-)-Ibotenic acid monohydrate
Health Hazard 2 GHS P Statement
Toxic if swallowed.
Molecular Formula C5H6N2O4·H2O
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.
MDL Number MFCD00149579
NMR conforms to structure
Infrared Spectrum Authentic
Health Hazard 1 Danger

Specifications

Chemical Name or Material (+/-)-Ibotenic acid monohydrate
CAS 60573-88-8
Assay ≥94% (HPLC)
Assay Percent Range 95%
Molecular Formula C5H6N2O4·H2O
Formula Weight 176.1
MDL Number MFCD00149579
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician.

GHS P Statement
Toxic if swallowed.

SDS & Safety Documents