missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ibotenic acid hydrate, 98+%

Potent NMDA and glutamate receptor agonist

$183.11 - $183.11

Chemical Identifiers

CAS 2552-55-8
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD00069294
InChI Key IRJCBFDCFXCWGO-UHFFFAOYSA-N
Synonym +/--ibotenic acid, 2-amino-2-3-oxo-1,2-oxazol-5-yl acetic acid, 4-isoxazoline-5-acetic acid, alpha-amino-3-oxo, 5-isoxazoleacetic acid, alpha-amino-2,3-dihydro-3-oxo, alpha-amino-2,3-dihydro-3-oxo-5-isoxazoleacetic acid, alpha-amino-3-hydroxy-5-isooxazolacetic acid, alpha-amino-3-hydroxy-5-isoxazoleacetic acid, amanita muscaria, ibotenate, ibotenic acid
PubChem CID 1233
ChEBI CHEBI:5854
IUPAC Name 2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid
SMILES C1=C(ONC1=O)C(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60748MCR View Documents Alfa Aesar™
J60748MCR
1mg Each for $183.11
 
Specifications

Chemical Identifiers

2552-55-8
158.113
IRJCBFDCFXCWGO-UHFFFAOYSA-N
1233
2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid
C5H6N2O4
MFCD00069294
+/--ibotenic acid, 2-amino-2-3-oxo-1,2-oxazol-5-yl acetic acid, 4-isoxazoline-5-acetic acid, alpha-amino-3-oxo, 5-isoxazoleacetic acid, alpha-amino-2,3-dihydro-3-oxo, alpha-amino-2,3-dihydro-3-oxo-5-isoxazoleacetic acid, alpha-amino-3-hydroxy-5-isooxazolacetic acid, alpha-amino-3-hydroxy-5-isoxazoleacetic acid, amanita muscaria, ibotenate, ibotenic acid
CHEBI:5854
C1=C(ONC1=O)C(C(=O)O)N

Specifications

White
2552-55-8
MFCD00069294
14,4877
IRJCBFDCFXCWGO-UHFFFAOYSA-N
2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid
1233
158.11 (Anhydrous)
≥98%
1mg
C5H6N2O4
UN3462
+/--ibotenic acid, 2-amino-2-3-oxo-1,2-oxazol-5-yl acetic acid, 4-isoxazoline-5-acetic acid, alpha-amino-3-oxo, 5-isoxazoleacetic acid, alpha-amino-2,3-dihydro-3-oxo, alpha-amino-2,3-dihydro-3-oxo-5-isoxazoleacetic acid, alpha-amino-3-hydroxy-5-isooxazolacetic acid, alpha-amino-3-hydroxy-5-isoxazoleacetic acid, amanita muscaria, ibotenate, ibotenic acid
C1=C(ONC1=O)C(C(=O)O)N
158.113
CHEBI:5854
Powder
Ibotenic acid hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

RTECSNumber : NY2100000