Please login to your online account to display your discounted pricing
IBI Scientific Hi/Lo Rocker Rocking Platform Shaker Accessory, Platforms
Add up to three additional platforms for increased capacity
$564.33 - $1,264.60
Specifications
For Use With Hi/Lo Profile Rocker
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
15-453-223
ibi scientific
ACCRC00P2
Plus 2 platform assembly Each for $914.38
15-453-222
ibi scientific
ACCRC00P1
Plus 1 platform assembly Each for $564.33
15-453-224
ibi scientific
ACCRC00P3
Plus 3 platform assembly Each for $1,264.60
Description & Specifications
Specifications
For Use With Hi/Lo Profile Rocker
Material Stainless Steel
  • Distance between platforms: 4.5 in.H (11cm)
  • Constructed of high-quality stainless steel