Please login to your online account to display your discounted pricing

IBC 293, Tocris

Selective agonist for the GPR109B (HM74) receptor; Antilipolytic

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material IBC 293
Assay Percent Range >99%
Molecular Formula C10H11N3O2
Formula Weight 205.22
Synonym 1-(1-Methylethyl)-1H-benzotriazole-5-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

246910

 
r & d systems
246910
10mg Each for $149.90

246950

 
r & d systems
246950
50mg Each for $632.23
Description & Specifications

Specifications

Formula Weight 205.22
Quantity 10mg
Chemical Name or Material IBC 293
Assay Percent Range >99%
Molecular Formula C10H11N3O2
CAS 306935-41-1
Synonym 1-(1-Methylethyl)-1H-benzotriazole-5-carboxylic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature