CBS Scientific Optimizer PCR Workstation with Stainless Steel Work Surface
Specially designed to protect against PCR reaction contamination.
Manufacturer:  CBS Scientific P04802SS
 View more versions of this product
Catalog No. S07079
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Prior UV irradiation protects against cross or airborne contamination in DNA sequencing and limits exposure to open-lab environment
  • Ample interior work space for PCR reaction supplies and equipment
  • Stainless-steel work surface and ceiling angle reflector for UV reflection
  • Hinged safety glass door for easy access and cleaning
  • Country-specific electrical outlet and two fluorescent lamps mounted in ceiling for work space visibility
  • 254nm UV germicidal lamp (with single or dual bulb) for increased UV dose with 12-hour countdown timer
  • Single or double access panel closure that closes during UV irradiation and slides into storage compartment during experiments
  • Available in three sizes
Specifications
Specifications
To protect against contamination in sensitive PCR amplification reactions
Stainless Steel
61cm
61cm
110V
121.92cm
Stainless Steel or black Formica working surface angle reflector, hinged Glass door, single panel closer, 2 fluorescent lamps, electric outlet, single bulb UV light with 12 hour timer, 110V duplex outlet
24 in.
24 in.
PCR Workstation
48 in.
Documents