missing translation for 'onlineSavingsMsg'
Learn More

Hypoxanthine 98.0+%, TCI America™

Supplier:  TCI America H03115G

 View more versions of this product

Catalog No. H03115G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hypoxanthine
C5H4N4O
5 g
FDGQSTZJBFJUBT-UHFFFAOYSA-N
6,7-dihydro-3H-purin-6-one
790
136.11
Crystalline Powder
68-94-0
MFCD00005725
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
O=C1N=CNC2=C1NC=N2
136.11
CHEBI:17368
≥98.0% (HPLC,E)

Chemical Identifiers

68-94-0
136.11
FDGQSTZJBFJUBT-UHFFFAOYSA-N
790
6,7-dihydro-3H-purin-6-one
C5H4N4O
MFCD00005725
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
CHEBI:17368
O=C1N=CNC2=C1NC=N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UP0791000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions