missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hypoxanthine, 99.5%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 122010250

 View more versions of this product

Catalog No. AC122010250



Description

Description

Specifications

Specifications

Hypoxanthine
0.5% max.
0.2% max.
Brown to Colorless
25g
99.4% min. (HPLC)
MFCD00005725
15, 4908
Solubility in water: practically insoluble. Other solubilities: soluble in diluted alkalies and acids, practically insoluble in most common organic, solvents
C1=NC2=C(N1)C(=O)N=CN2
136.11
CHEBI:17368
Liquid
Authentic
Glass bottle
95% min. (c=0.1, 0.2M NaOH, 430nm) 1cm cell
>250.0°C
68-94-0
C5H4N4O
26, 416
3h-purin-6-ol, 6-hydroxypurine, 6-oxopurine, 7h-purin-6-ol, 9h-purin-6-ol, hypoxanthine, purin-6 1h-one, sarcine, sarkin, sarkine
FDGQSTZJBFJUBT-UHFFFAOYSA-N
3,7-dihydropurin-6-one
790
136.11
99.5%

Chemical Identifiers

68-94-0
136.11
FDGQSTZJBFJUBT-UHFFFAOYSA-N
790
3,7-dihydropurin-6-one
C5H4N4O
MFCD00005725
3h-purin-6-ol, 6-hydroxypurine, 6-oxopurine, 7h-purin-6-ol, 9h-purin-6-ol, hypoxanthine, purin-6 1h-one, sarcine, sarkin, sarkine
CHEBI:17368
C1=NC2=C(N1)C(=O)N=CN2
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-697-3

SDS Documents