Please login to your online account to display your discounted pricing

Thermo Scientific™ Hypersil™ MOS C8 LC Columns


Reliably analyze moderately hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.

Manufacturer: thermo scientific™  30205014001

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-150-545

  • / Pack of 4 Each
 

Description & Specifications

Specifications

Column Type Reversed Phase
Max. Pressure 400bar
Temperature 60°C
Column Format Guard Cartridge, 4 PK
Diameter (Metric) 4mm
Packing Material ----
Particle Size 5µm
Pore Size 120Å
pH 2 to 8
For Use With Uniguard Direct-connection and Stand Alone Holder
Stationary Phase ----
Surface Area 170m²/g
Length (Metric) 10mm
USP Type L7
Endcapped -
Diameter (Metric) 4mm
Pore Size 120Å
Carbon Load -
Item Description 4.0 or 4.6mm ID; 10mm length; 5μm particle size

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • The 10mm design offers maximum protection with minimal increase in retention Fit Thermo Scientific UNIGUARD direct connection and stand alone holders
  • Available in packs of 4

Feature of MOS (C8) Solid Phase

  • Reliable, less retentive phase