Please login to your online account to display your discounted pricing

Thermo Scientific™ Hypersil™ MOS C8 LC Columns


Reliably analyze moderately hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.

Manufacturer: thermo scientific™  30205104630

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-150-549

  • / Each
 

Description & Specifications

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6mm
Carbon Load 6.5%
Column Type Reversed Phase
Stationary Phase C8
For Use With ----
Surface Area 170m²/g
Pore Size 120Å
Max. Pressure 400bar
Particle Size 5µm
Temperature 60°C
Length (Metric) 100mm
pH 2 to 8
USP Type L7
Packing Material Spherical, fully porous Type A silica
Endcapped No
Certifications/Compliance Manufactured under ISO 9001-2000 quality guidelines.

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable

Ordering Information:

Other column dimensions are available. Also available in 10μm. Please call Customer Service for more information.