Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ Silica Analytical HPLC Columns, 3µm
Separate nonpolar and moderately polar organic compounds by normal phase chromatography using Thermo Scientific™ Hypersil GOLD™ Silica Analytical HPLC Columns.
$1,169.18 - $1,169.18
Products
Catalog Number Mfr. No. Diameter Length (Metric) Price Quantity    
25-103-154030
thermo scientific™
25103154030
Each for $1,169.18
Description & Specifications
Achieve outstanding peak shape and sensitivity with Thermo Scientific™ Hypersil GOLD™ Silica Analytical HPLC Columns. The unbonded, highly pure, base-deactivated silica media efficiently separates nonpolar and moderately polar organic compounds by normal phase chromatography. Available 1.9µm particle size columns can improve the speed and efficiency of your applications.
About the Hypersil GOLD Family of HPLC Columns:
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5µm particle sizes for all your separation needs