Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ C4 Analytical HPLC Columns, 3µm
Achieve faster separations for very hydrophobic analytes with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns.
$1,204.88 - $1,204.88
Products
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Particle Size Price Quantity    
25-503-051030
thermo scientific™
25503051030
1mm 50mm 3µm Each for $1,204.88
Description & Specifications
Feature lower hydrophobicity than C18 or C8 and are recommended for very hydrophobic analytes with Thermo Scientific™ Hypersil GOLD™ 3μm C4 Analytical HPLC Columns. Based on high purity silica, Hypersil GOLD is a state-of-the-art family of columns designed to meet the challenges of modern chromatography.
Similar Selectivity, Less Retention Than C18 or C8 Columns
  • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
  • Ideal for analysis of hydrophobic polypeptides and proteins
Superior Performance
  • Highly symmetrical peak shapes
  • 1.9µm particle size columns improve speed and efficiency
About the Hypersil GOLD Family of HPLC Columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5µm particle sizes for all your separation needs