Please login to your online account to display your discounted pricing
Thermo Scientific™ Hypersil GOLD™ AX Drop-In Guard Cartridges
Protect Thermo Scientific™ Hypersil GOLD™ AX Analytical HPLC Columns from sample debris and interfering substances, economically.
$572.06 - $572.06
Products
Catalog Number Mfr. No. Diameter Length (Metric) Particle Size Price Quantity    
26-105-011001
thermo scientific™
26105011001
Pack of 4 Each for $572.06
Description & Specifications
Protect your analytical HPLC column from damage caused by sample debris and interfering substances. It's the economical way to extend the lifetime of your analytical column with minimal impact on performance.
Use Thermo Scientific™ Hypersil GOLD™ AX Drop-In Guard Cartridges with Hypersil GOLD AX Analytical HPLC columns. Select the guard column that best matches your analytical column I.D. and particle size. These convenient, drop-in cartridges are simple to install and replace using the Thermo Scientific™ UNIGUARD™ Direct-Connection Guard Cartridge Holder or a stand-alone holder.