Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Manufacturer:  Thermo Scientific™ 60104234
 View more versions of this product
Catalog No. 03-251-254
Specifications
Description
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
Description & Specifications
Specifications
48 Position Extraction Plate Plugs
Vacuum Manifolds