Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 1-10µL
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Manufacturer: thermo scientific™  60109201
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 03-250-450
  • / Pack of 96 Each
Description & Specifications
Specifications
Item Description BioBasic C18, 1 to 10µL Volume
Particle Size 40 to 60µm
Description BioBasic C18, 1 to 10µL Volume
For Use With (Equipment) Microscale Solid Phase Extraction
For Use With Microscale Solid Phase Extraction
Sorbent BioBasic C18
Type Tip
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.