missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HyperSep™ Retain PEP Cartridges

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.

Manufacturer:  Thermo Scientific™ 60107202

 View more versions of this product

Catalog No. 03-150-634


 

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Features:
Ideal for large samples, Hypersep Retain SPE cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • Available in a wide range of chemistries and bed weights
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
Description & Specifications

Specifications

30mg bed weight; 3mL column volume
Recovery for Polar and Non-polar Analytes
Extraction Column
30mg
Recovery for Polar and Non-polar Analytes
HyperSep Retain PEP
40 - 60µm
3mL