missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HyperSep™ Polypropylene Lab Plates

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Manufacturer:  Thermo Scientific™ 60110307

 View more versions of this product

Catalog No. 03-250-569



Description

Description

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polypropylene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include: 

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
Specifications

Specifications

Titanium Dioxide; Polypropylene
HyperSep
96-Well Plate
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep Titanium Dioxide
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.