Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Glass Blocks for Vacuum Manifold
Order the glass block, which is the vacuum chamber of this manifold, if you need a replacement.
$889.20 - $889.20
Specifications
For Use With Glass Block Vacuum Manifold
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    
60-104-254
thermo scientific™
60104254
Glass Block, 24 Port Vacuum Manifold, 1pk Each for $889.20
Description & Specifications
Specifications
For Use With Glass Block Vacuum Manifold
Order the Thermo Scientific™ HyperSep™ Vacuum Manifold Glass Block if you need a replacement. The vacuum chamber for this manifold is a glass block that allows you to see the extraction process and provides easy cleaning. The manifold enables sample preparation using HyperSep solid phase extraction (SPE) cartridges.