Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Stopcocks
Use this stopcock to control the flow of extract exiting your SPE cartridge on the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold.
Manufacturer: thermo scientific™  60104242
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 03-150-622
  • / Pack of 16 Each
Description & Specifications
Specifications
For Use With Glass Block Vacuum Manifold
Description Stopcocks for 24 port Glass Block Vacuum Manifold, 16Pk
Control the flow of extract exiting your SPE cartridge using Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Stopcocks. The manifold enables you to process 16 or 24 samples through HyperSep SPE solid phase extraction cartridges under vacuum. Stopcocks are attached to the tip of cartridge for manual control of flow.