Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE multipacks for QuEChERS extractions.
Manufacturer: thermo scientific™  60105333
 View more versions of this product
Catalog No. 03-052-746
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) QuEChERS Method
For Use With QuEChERS Method
Item Description 4000mg Magnesium Sulfate, 1000mg Sodium Chloride, 500mg Sodium Citrate dibasic sesquihydrate, 1000mg sodium citrate tribasic, 50pk
Type QuEChERS Sample Prep
Sorbent Magnesium Sulfate, Sodium Chloride, Sodium Citrate Dibasic Sesquihydrate, Sodium Citrate Tribasic
Description 4000mg Magnesium Sulfate, 1000mg Sodium Chloride, 500mg Sodium Citrate dibasic sesquihydrate, 1000mg sodium citrate tribasic, 50pk
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Convenient Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks each include 50 metallized pouches of pre-prepared sorbent, along with 50 empty centrifuge tubes with plug seal caps.