Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE multipacks for QuEChERS extractions.
Manufacturer:  Thermo Scientific™ 60105336
 View more versions of this product
Catalog No. 03-052-743
Specifications
Description
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Convenient Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks each include 50 metallized pouches of pre-prepared sorbent, along with 50 empty centrifuge tubes with plug seal caps.
Description & Specifications
Specifications
1500mg Sodium Chloride, 6000mg Magnesium Sulfate, 50pk
QuEChERS Method
QuEChERS Sample Prep
QuEChERS Method
Sodium Chloride, Magnesium Sulfate
1500mg Sodium Chloride, 6000mg Magnesium Sulfate, 50pk