Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE multipacks for QuEChERS extractions.
Manufacturer: thermo scientific™  60105337
 View more versions of this product
Catalog No. 03-052-741
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) QuEChERS Method
For Use With QuEChERS Method
Sorbent Magnesium Sulfate, NaCl, Sodium Citrate Dihydrate, Disodium Citrate Sesquihydrate
Item Description 6000mg Magnesium Sulfate, 1500mg NaCl, 1500mg sodium citrate dihydrate, 750mg disodium citrate sesquihydrate
Type QuEChERS Sample Prep
Description 6000mg Magnesium Sulfate, 1500mg NaCl, 1500mg sodium citrate dihydrate, 750mg disodium citrate sesquihydrate
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Convenient Thermo Scientific™ HyperSep™ Dispersive SPE Multipacks each include 50 metallized pouches of pre-prepared sorbent, along with 50 empty centrifuge tubes with plug seal caps.