Please login to your online account to display your discounted pricing

Thermo Scientific™ HyperSep™ Diol Plates


Obtain excellent normal phase extraction of polar compounds with these diol SPE well plates and individual wells.

Manufacturer: thermo scientific™  60300630

 View more versions of this product

Catalog No. 03-251-481

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Extraction of Polar Compounds
Particle Size 40 - 60µm
For Use With Extraction and Purification of Polar Compounds
Sorbent HyperSep Diol
Item Description Diol 10mg/1mL 96 Removable Well Plate, 1pk
Type 96-well Plate
Description Diol 10mg/1mL 96 Removable Well Plate, 1pk

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

The sorbent used in Thermo Scientific™ HyperSep™ Diol Plates features short alkyl chains with polar functional groups, and are ideal for the isolation of polar compounds. Typical applications include:

  • Normal phase extraction of polar compounds
  • Purification of polar compounds

HyperSep Diol SPE 96-Well Plates and Individual Wells are typically used as an alternative to silica in normal phase applications where silica’s acidic character can be problematic.

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.