Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Cyano Plates
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE well plates and individual wells.
Manufacturer: thermo scientific™  60300823
 View more versions of this product
Catalog No. 03-052-238
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) Retention of Polar Compounds from Non-polar Matrices
Particle Size 40 - 60µm
Sorbent HyperSep Cyano
Type 96-well Plate
For Use With Retention of Polar Compounds from Non-polar Matrices
Item Description Cyano 50mg/1mL 96 Removable Well Plate, 1pk
Description Cyano 50mg/1mL 96 Removable Well Plate, 1pk
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano Plates are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
  • Retention of polar compounds from hexane and oils
  • Reversed phase extraction of moderately polar compounds
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.