Please login to your online account to display your discounted pricing
Thermo Scientific™ HyperSep™ Aminopropyl Plates
Obtain excellent retention of drugs, metabolites and structural isomers with these aminopropyl SPE well plates that allow both polar and anion exchange interactions.
Manufacturer: thermo scientific™  60300507
 View more versions of this product
Catalog No. 03-251-353
  • / Pack
Description & Specifications
Specifications
For Use With (Equipment) Separation of Structural Isomers, Drugs and Metabolites in Biological Fluids
For Use With Separation of Structural Isomers, Drugs and Metabolites in Biological Fluids
Sorbent HyperSep Aminopropyl
Type 96-well Plate
Particle Size 40 - 60µm
Item Description Aminopropyl 50mg/1mL 96 Removable Well Plate, 1pk
Description Aminopropyl 50mg/1mL 96 Removable Well Plate, 1pk
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ Aminopropyl Plates feature a polar sorbent that allows both polar and anion exchange interactions. The chemistry offers excellent retention of drugs, metabolites and structural isomers. Typical applications include:
  • Separation of structural isomers
  • Drugs and metabolites in biological fluids
  • Saccharides, phenols and petroleum products
The sorbent used in HyperSep Aminopropyl SPE 96-Well Plates and Individual Wells acts as a weak anion exchanger, and also provides enhanced retention of strong anions like sulfonic acids.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.