missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HyPerforma™ Drum Accessories

Enable easy transport of BioProcess Container Systems™ (BPC™s) with Thermo Scientific™ HyPerforma™ Drum Accessories.

$278.48 - $278.48

Specifications

Capacity (Metric) 200L
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
SV5002903 Thermo Scientific™
SV5002903
Plastic Drum Dolly Each for $278.48
Specifications

Description

Enable easy transport of BioProcess Container Systems™ (BPC™s) with Thermo Scientific™ HyPerforma™ Drum Accessories.

Support Plate
  • Round plastic disk for use with conical HDPE drums with 4 ″ port.
  • Supports up to 200L drums

Plastic Drum Dolly
  • Holds conical and flat-bottom drums
  • Supports up to 200L drums

  • Ordering Alerts:

    Dimensions, Plastic Drum Dolly: 24.38 Dia. × 7.13 in. H

    Description & Specifications

    Specifications

    200L