Learn More
Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals
$116.55 - $1118.01
Chemical Identifiers
CAS | 83-49-8 |
---|---|
Molecular Formula | C24H40O4 |
Molecular Weight (g/mol) | 392.58 |
MDL Number | MFCD00003681 |
InChI Key | DGABKXLVXPYZII-SIBKNCMHSA-N |
Synonym | hyodeoxycholic acid |
PubChem CID | 124303820 |
IUPAC Name | (4S)-4-[(3S,5S,6R,8R,9R,10S,13S,14R,17S)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
SMILES | [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2050606
|
Thermo Scientific Chemicals
B2050606 |
5 g |
Each for $116.55
|
|
|||||
AAB2050614
|
Thermo Scientific Chemicals
B2050614 |
25 g |
Each for $347.79
|
|
|||||
AAB2050622
|
Thermo Scientific Chemicals
B2050622 |
100 g |
Each for $1,118.01
|
|
|||||
Description
It is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.
Solubility
Solubility in methanol, very faint turbidity. Slightly soluble in alcohol, acetone, ether and very slightly soluble in chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
83-49-8 | |
392.58 | |
DGABKXLVXPYZII-SIBKNCMHSA-N | |
124303820 | |
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C |
C24H40O4 | |
MFCD00003681 | |
hyodeoxycholic acid | |
(4S)-4-[(3S,5S,6R,8R,9R,10S,13S,14R,17S)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid |
Specifications
83-49-8 | |
C24H40O4 | |
5 g | |
hyodeoxycholic acid | |
+8° (c=2 in Ethanol) | |
(4S)-4-[(3S,5S,6R,8R,9R,10S,13S,14R,17S)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid | |
124303820 | |
96% |
197°C to 199°C | |
MFCD00003681 | |
14,4857 | |
DGABKXLVXPYZII-SIBKNCMHSA-N | |
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C | |
392.58 | |
392.59 | |
Hyodeoxycholic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-483-2
RTECSNumber : FZ2050000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only