Hygiena SnapShot™ ATP Devices
Sensitive, accurate, and reliable single-shot device
$524.81 - $524.81
Specifications
Storage Requirements 2° to 8°C
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
22-037-757 View Documents Hygiena
CH1616
Charm LUM-T, Firefly luminometers Pack of 100 Each for $524.81
Case of 5 Pack for $2,624.05
Description
Description
Snapshot is designed to replicate the ATP reaction by generating the same light output as the device it is replacing, therefore eliminating the need to change current pass/fail limit settings on the luminometer. In addition, Snapshot uses Hygiena's unique liquid-stable reagent giving it more accurate and consistent results.
  • Consistent, reproducible results
  • No need to change Pass/Fail setting - Snapshot works exactly the same as the device it is replacing
  • Patented Snap-Valve™ technology and self-contained liquid stable reagent minimizes costs and maximizes performance
  • Premoistened swab bud
  • Write-on swab label
  • Unprecedented accuracy and reproducibility
Specifications
Specifications
2° to 8°C