Ansell HyFlex™ 11-931 Cut-Resistant Lightweight Gloves, Palm Coated
Lightest weight cut-resistant and oil-repellent gloves with reinforced thumb crotch that provides up to 12x durability for extended wear.
$201.29 - $201.29
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Coating Material Nitrile foam
Color Black
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-010-278
View Documents Promotion Details
Ansell
11931080
8
Pack of 12 Each for $201.29
Case of 12 Pack for $2,415.48
19-010-279
View Documents Promotion Details
Ansell
11931090
9
Pack of 12 Each for $201.29
Case of 12 Pack for $2,415.48
19-010-280
View Documents Promotion Details
Ansell
11931100
10
Pack of 12 Each for $201.29
Case of 12 Pack for $2,415.48
19-010-281
View Documents Promotion Details
Ansell
11931110
11
Pack of 12 Each for $201.29
Case of 12 Pack for $2,415.48
Description
Description
  • RIPEL™ technology keeps oil from making contact with skin, keeping hands safe and dry
  • Provides protection from cuts and oil exposure, from light to heavy, across multiple applications
  • Provides dexterity and high level of comfort with ZONZ technology
  • ANSELL GRIP™ Technology for safe handling in dry-to-oily applications
  • FORTIX Technology for extended product use life and increased protection with up to 12x durability in thumb crotch
  • Ultra-lightweight 18 gauge design provides dexterity and high level of comfort
  • Protects against lacerations, oil and industrial fluid exposure
Specifications
Specifications
ANSI 4
Knit
Black
Cut Resistant Gloves
Inspection, assembly, fastening components, equipment repair and maintenance, automotive applications, press shop operations
Dyneema™ Diamond Technology
HyFlex™ 11-391
8.3 in.
CE, EN 420:2003 + A1:2009; Category II
Nitrile foam
ANSI A2
EN 388:4X42B
Ansell GRIP™ Technology
ISO 188889
Reinforced
21.2 cm