Ansell HyFlex™ 11-435 Cut-Resistant Glove, Palm Coated Polyurethane and Nitrile
Knit gloves with palm-coated finishing provide extreme resistance to cuts and burrs while providing a comfortable fit, flexibility and cool feeling.
$183.15 - $189.20
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category ll
Construction Knit
Coating Material Nitrile
Color Black,Gray
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-010-215
View Documents Promotion Details
Ansell
288904
6
Dozen of 12 Each for $183.15
Case of 12 Dozen for $2,197.80
19-010-216
View Documents Promotion Details
Ansell
288905
7
Dozen of 12 Each for $183.15
Case of 12 Dozen for $2,197.80
19-010-217
View Documents Promotion Details
Ansell
288906
8
Dozen of 12 Each for $183.15
Case of 12 Dozen for $2,197.80
19-010-220
View Documents Promotion Details
Ansell
288909
11
Dozen of 12 Each for $189.20
Case of 12 Dozen for $2,270.40
Description
Description
  • Unique combination of Dyneema, Nylon, Lycra, and glass fiber
  • ISO Cut level C
  • Water-based polyurethane and NBR
  • Ultra strong fibers provide extreme resistance to cuts and burrs
  • Anti-static for improved safety in critical areas
  • Silicone free
  • Oeko-Tex 100 approved
  • Also available in vend pack
Specifications
Specifications
ANSI 5
Knit
Black,Gray
Specific
Cut Resistant Gloves
Handling metal sheets and panels, handling sharp and rough edges, body painting, mounting and dismantling, foundry operations, maintenance
Nylon, Lycra™, Fiberglass, Dyneema™
8.8 in.
CE, EN 420:2003 + A1:2009; Category ll
Nitrile
ANSI A3
Yes
EN 388:4542; EN 388:4X42C
Knit
HyFlex™ 11-435
22.5 cm