Please login to your online account to display your discounted pricing

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412152500

 View more versions of this product

Catalog No. AC412152500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.0% (as mixture of isomers)
Formula Weight 144.17
Physical Form Liquid
Quantity 250g
Color Clear
TSCA TSCA
EINECS Number 213-090-3
RTECS Number OZ4750000
Density 1.0300g/mL
CAS Max % 100.0
CAS Min % 97.0
Stabilizer 150 to 300ppm MEHQ
Water 0.1% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
CAS 27813-02-1
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 1 Warning
MDL Number MFCD00004536
Linear Formula CH2=C(CH3)CO2CH2CH(OH)CH3
Molecular Formula C7H12O3
Name Note 96.5%
Chemical Name or Material Hydroxypropyl methacrylate
Assay Percent Range 97+%

Specifications

Chemical Name or Material Hydroxypropyl methacrylate
Name Note 96.5%
CAS 27813-02-1
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0% (as mixture of isomers)
Assay Percent Range 97+%
Molecular Formula C7H12O3
Linear Formula CH2=C(CH3)CO2CH2CH(OH)CH3
Formula Weight 144.17
MDL Number MFCD00004536
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.

SDS & Safety Documents