Please login to your online account to display your discounted pricing

Hydroxypropyl-β-cyclodextrin, 97%, ACROS Organics™


Quantity 100g
Packaging Glass Bottle

Manufacturer: fisher scientific acros  297561000

 View more versions of this product

Catalog No. AC297561000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (on dry substance), 2.0% max. residual beta-cyclodextrin (on dry substance)
Physical Form Powder
Quantity 100g
Packaging Glass Bottle
Melting Point 278.0°C
Color White to slightly yellow
Chemical Name or Material Hydroxypropyl-β-cyclodextrin
NMR (degree of substitution) 2 to 6 substituents are located mainly at O(2) of the anhydroglucopyranose units
Water 7.5% max. (K.F.)
MDL Number MFCD16621721
Assay Percent Range 97%
Infrared Spectrum Authentic
CAS 128446-35-5

Specifications

Chemical Name or Material Hydroxypropyl-β-cyclodextrin
Assay ≥96.0% (on dry substance), 2.0% max. residual beta-cyclodextrin (on dry substance)
Assay Percent Range 97%
MDL Number MFCD16621721