missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydroxypropyl cellulose, M.W. 100,000

Thickener, binder, and emulsifier

Manufacturer:  Alfa Aesar™ 4340036

 View more versions of this product

Catalog No. AA4340036



Specifications

Specifications

Hydroxypropyl cellulose
500g
M.W. 100,000
(C24H44O16)n
14,4841
Soluble in polar organic solvents, in water at room temperature
Odorless
7
9004-64-2
MFCD00132688
4-1-aminopropyl-n,n,3-trimethyl-aniline
Powder

Chemical Identifiers

9004-64-2
(C24H44O16)n
MFCD00132688
4-1-aminopropyl-n,n,3-trimethyl-aniline
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents