Please login to your online account to display your discounted pricing

Hydroxymethylpolystyrene resin, 1% crossl. with DVB, 1.0 to 1.2mmol/g, 100 to 200 mesh

Chemical Identifiers

Chemical Name or Material Hydroxymethylpolystyrene resin
CAS 66072-40-0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC349510050

  SDS
thermo fisher scientific
349510050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Hydroxymethylpolystyrene resin
Packaging Glass bottle
CAS 66072-40-0

For solid support synthesis